Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A Closer Look at Polymers Lab Investigation
SCI_ED

Catalog No. S67401
Change view
Click to view available options
A Closer Look at Polymers Lab Investigation
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S67401 A Closer Look at Polymers Lab Investigation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S67401 Supplier Neo/Sci Corporation Supplier No. 201753
This item is not returnable. View return policy

Make and compare Slime and Flexi-Putty.

  • Gain an understanding of long-chain polymers and cross-linked agents
  • Develop an appreciation for the chemistry behind these important polymers
  • Learn about their uses in everyday materials
  • Have fun with the experimental products

Order Info

Class size: 40 students.

Specifications

For Use With (Application) Gain an understanding of long-chain polymers and cross-linked agents, learn about their uses in everyday materials
Class Size 40 Students
Product Type A Closer Look at Polymers Lab Investigation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.