Learn More
A Closer Look at Pond Life Lab Investigation
SCI_ED
Catalog No. S23199
Change view
Click to view available options
Includes:
Pond life lab investigation kit
Catalog No. Includes
S23199 Pond life lab investigation kit
1 options
This item is not returnable. View return policy
Catalog No. S23199 Supplier Neo/Sci Corporation Supplier No. 201233
This item is not returnable. View return policy
Includes
  • Dried pond mix
  • Clear pond containers
  • Protist-slowing agent
  • Test vials
  • Pipets
  • Vital stain
  • Microscope slides
  • Simulated pollutants
  • Comprehensive teacher and student guides
  • Illustrated, dichotomous key to microlife forms
Observe microbial populations and learn to calculate population diversity indices.
  • Investigate a unique pond ecosystem
  • Build a miniature pond ecosystem
  • Explore the diverse life forms that evolve
  • Monitor microbial populations
  • Gain experience in calculating population diversity indices
  • Design unique experiments to determine the population dynamics within a micro-ecosystem as well as the effects of various pollutants
Order Info
Class size: 40 students.
Specifications
Includes Pond life lab investigation kit
Product Type A Closer Look at Pond Life Lab Investigation