Learn More
Selleck Chemical LLC a-cyano-4-hydroxycinnamic acid
SDP
Catalog No. 5000001679
Encompass_Preferred
4-Chloro- -cyanocinnamic acid ( -CHCA) is a classic monocarboxylate transporters (MCT) inhibitor -cyano-4-hydroxycinnamate (CHC) has a 10-fold selectivity for MCT1 compared to other MCTs
Catalog No. 50-000-01679 Supplier Selleck Chemical LLC Supplier No. S86121G
4-Chloro- -cyanocinnamic acid ( -CHCA) is a classic monocarboxylate transporters (MCT) inhibitor -cyano-4-hydroxycinnamate (CHC) has a 10-fold selectivity for MCT1 compared to other MCTs