Learn More
Alkali Scientific a Cyclodextrin, 1 G
SDP
Catalog No. 5000660642
Encompass_Preferred
Alpha Cyclodextrin (1g) is a cyclic oligosaccharide used in laboratory applications. It is commonly employed in various chemical and biochemical procedures where its unique molecular structure plays a crucial role in forming inclusion complexes with various organic and inorganic molecules. Cyclodextrins like alpha-cyclodextrin are widely utilized in the pharmaceutical industry for drug delivery systems, in food processing to improve solubility, and as a chiral selector in chromatography. It can also be used in research applications related to enzymology and molecular recognition. This specific formulation, 1g in size, is suitable for precise laboratory experiments requiring high-quality reagents.
Catalog No. 50-006-60642 Supplier Alkali Scientific Supplier No. ACY001CP
Add to Cart
Add to Cart
Alpha Cyclodextrin (1g) is a cyclic oligosaccharide used in laboratory applications. It is commonly employed in various chemical and biochemical procedures where its unique molecular structure plays a crucial role in forming inclusion complexes with various organic and inorganic molecules. Cyclodextrins like alpha-cyclodextrin are widely utilized in the pharmaceutical industry for drug delivery systems, in food processing to improve solubility, and as a chiral selector in chromatography. It can also be used in research applications related to enzymology and molecular recognition. This specific formulation, 1g in size, is suitable for precise laboratory experiments requiring high-quality reagents.