Learn More
ALADDIN SCIENTIFIC CORPORATION a-Cyclodextrin, 100g
Catalog No. 5000082547
Encompass_Preferred
a-Cyclodextrin can be used- In fabricating reversible supramolecular assemblies As a modifier to enhance the solubility of oleic acid stabilized nanoparticles In the preparation of polyrotaxanes with ply(ethylene glycol) In the synthesis of cationic star polymers with oligoethylenimine
Catalog No. 50-000-82547 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. C106777100G
a-Cyclodextrin can be used- In fabricating reversible supramolecular assemblies As a modifier to enhance the solubility of oleic acid stabilized nanoparticles In the preparation of polyrotaxanes with ply(ethylene glycol) In the synthesis of cationic star polymers with oligoethylenimine