Learn More
Alkali Scientific a Cyclodextrin, 25 G
SDP
Catalog No. 5000660150
Encompass_Preferred
Alpha Cyclodextrin (25g) is a cyclic oligosaccharide widely used in laboratory research and applications. Its unique molecular structure allows it to form inclusion complexes with a range of molecules, making it essential in various chemical processes, particularly in the pharmaceutical and food industries. This 25g reagent is commonly used in drug formulation, chromatography for chiral separations, and as a solubility enhancer in aqueous solutions. In biochemical applications, alpha-cyclodextrin is used to study molecular interactions, enzyme-substrate reactions, and as a stabilizing agent for various compounds. This reagent's versatile use makes it invaluable in analytical, chemical, and industrial laboratories.
Catalog No. 50-006-60150 Supplier Alkali Scientific Supplier No. ACY002CP
Alpha Cyclodextrin (25g) is a cyclic oligosaccharide widely used in laboratory research and applications. Its unique molecular structure allows it to form inclusion complexes with a range of molecules, making it essential in various chemical processes, particularly in the pharmaceutical and food industries. This 25g reagent is commonly used in drug formulation, chromatography for chiral separations, and as a solubility enhancer in aqueous solutions. In biochemical applications, alpha-cyclodextrin is used to study molecular interactions, enzyme-substrate reactions, and as a stabilizing agent for various compounds. This reagent's versatile use makes it invaluable in analytical, chemical, and industrial laboratories.