Learn More
Alkali Scientific a Cyclodextrin, 5 G
SDP
Catalog No. 5000660633
Encompass_Preferred
Alpha Cyclodextrin (5g) is a highly pure cyclic oligosaccharide that plays a crucial role in laboratory experiments, particularly in the study of molecular interactions and enzyme catalysis. Its unique ability to form inclusion complexes with various molecules makes it a valuable reagent in research. Alpha-cyclodextrin is widely used in chromatography for the separation of chiral compounds, as well as in the pharmaceutical industry to enhance the solubility and stability of drugs. This 5g formulation is ideal for small-scale laboratory work, offering precise amounts of high-quality cyclodextrin for various chemical and biochemical applications in a wide range of research fields.
Catalog No. 50-006-60633 Supplier Alkali Scientific Supplier No. ACY003CP
Add to Cart
Add to Cart
Alpha Cyclodextrin (5g) is a highly pure cyclic oligosaccharide that plays a crucial role in laboratory experiments, particularly in the study of molecular interactions and enzyme catalysis. Its unique ability to form inclusion complexes with various molecules makes it a valuable reagent in research. Alpha-cyclodextrin is widely used in chromatography for the separation of chiral compounds, as well as in the pharmaceutical industry to enhance the solubility and stability of drugs. This 5g formulation is ideal for small-scale laboratory work, offering precise amounts of high-quality cyclodextrin for various chemical and biochemical applications in a wide range of research fields.