Learn More
Cayman Chemical α-Cyclodextrin (hydrate), Purity: ≥95%, CAS: 51211-51-9, 50 g
SDP
Catalog No. 502989174
Encompass_Preferred
A cyclic hexasaccharide; forms inclusion complexes with wide variety of molecules, improving their solubility and stabilization; has commonly been used in the separation and purification of chemicals on an industrial scale; has also been used in the pharmaceutical and food industries
Catalog No. 50-298-9174 Supplier Cayman Chemical Supplier No. 3121950 G
A cyclic hexasaccharide; forms inclusion complexes with wide variety of molecules, improving their solubility and stabilization; has commonly been used in the separation and purification of chemicals on an industrial scale; has also been used in the pharmaceutical and food industries