Learn More
Cayman Chemical α-Cyano-4-hydroxycinnamic Acid, Purity: ≥98%, CAS: 28166-41-8, 5 g
SDP
Catalog No. 502972800
Encompass_Preferred
An MCT1 inhibitor; blocks pyruvate transport in rat heart mitochondria (IC50 = 1.5 μM); commonly used as a matrix to facilitate peptide ionization in MALDI-TOF mass spectrometry applications
Catalog No. 50-297-2800 Supplier Cayman Chemical Supplier No. 152545 G
An MCT1 inhibitor; blocks pyruvate transport in rat heart mitochondria (IC50 = 1.5 μM); commonly used as a matrix to facilitate peptide ionization in MALDI-TOF mass spectrometry applications