Learn More
Cayman Chemical a-D-Glucose-1-phosphatesodI 1g
SDP
Catalog No. 502991981
Encompass_Preferred
An intermediate in glycogen metabolism; precursor in the biosynthesis of UDP-glucose; can be formed during glycogen breakdown via phosphorolytic cleavage of glycogen by glycogen phosphorylase; can be converted to glucose-6-phosphate by phosphoglucomutase; is combined with CTP to form CDP-glucose in S. typhi
Catalog No. 50-299-1981 Supplier Cayman Chemical Supplier No. 305661 G
An intermediate in glycogen metabolism; precursor in the biosynthesis of UDP-glucose; can be formed during glycogen breakdown via phosphorolytic cleavage of glycogen by glycogen phosphorylase; can be converted to glucose-6-phosphate by phosphoglucomutase; is combined with CTP to form CDP-glucose in S. typhi