Learn More
Cayman Chemical α-D-Glucose-1-phosphate (sodium salt hydrate), Purity: ≥95%, CAS: 150399-99-8, 5 g
SDP
Catalog No. 502991275
Encompass_Preferred
An intermediate in glycogen metabolism; precursor in the biosynthesis of UDP-glucose; can be formed during glycogen breakdown via phosphorolytic cleavage of glycogen by glycogen phosphorylase; can be converted to glucose-6-phosphate by phosphoglucomutase; is combined with CTP to form CDP-glucose in S. typhi
Catalog No. 50-299-1275 Supplier Cayman Chemical Supplier No. 305665 G
An intermediate in glycogen metabolism; precursor in the biosynthesis of UDP-glucose; can be formed during glycogen breakdown via phosphorolytic cleavage of glycogen by glycogen phosphorylase; can be converted to glucose-6-phosphate by phosphoglucomutase; is combined with CTP to form CDP-glucose in S. typhi