Learn More
Thermo Scientific A/D Interface, 2 Channels
Supplier:  Thermo Scientific 7200.0005
Encompass
Dual channel USB data acquisition module with 16 bit resolution, 1 kS/s sampling rate, and analog inputs with software-selectable ranges converts analog voltage signals to digital data for general purpose measurement and data logging applications.
Catalog No. NC1200937