Learn More
Cayman Chemical a dIfluoromethylDLArgInIn 10mg
SDP
Catalog No. 502972296
Encompass_Preferred
An enzyme-activated, irreversible inhibitor of the arginine decarboxylases of E. coli (Ki = 800 µM), P. aeruginosa, and K. pneumoniae; prevents the osmotic stress-induced increase in arginine decarboxylase activity and putrescine synthesis in oat leaf cells at 0.01 mM
Catalog No. 50-297-2296 Supplier Cayman Chemical Supplier No. 1641510 MG
An enzyme-activated, irreversible inhibitor of the arginine decarboxylases of E. coli (Ki = 800 µM), P. aeruginosa, and K. pneumoniae; prevents the osmotic stress-induced increase in arginine decarboxylase activity and putrescine synthesis in oat leaf cells at 0.01 mM