Learn More
Cytiva A/E Glass Fiber Disc Filter - 1 µm, 76 mm (100/pkg)
Supplier:  Cytiva 61663
Encompass
Superior Grade Filters for a Variety of Biological and Environmental Methods-Type A/E meets the requirements for suspended solids testing, as described in Standard Methods for the Examination of Water and Wastewater, current edition.-Reduces filtration costs and premature clogging when filtering difficult-to-filter or highly contaminated solutions.-Extends filter life and reduces final filter changes with high capacity prefilters.-Eliminates sample contamination. Binder-free borosilicate glass fiber has no added extractables.-Filters a wide range of particulate loads and viscous solutions with a selection of filter thicknesses from which to choose.-Filters with binder offer excellent wet strength for easy handling and filter integrity. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3264