Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC a-Fluoromethylhistid 1mg | 81839-27-2 | 260.09 g/mol | C7H12Cl2FN3O2 | 1 MG
SDP

Catalog No. 5000307649
Encompass_Preferred

α-Fluoromethyl-L-histidine dihydrochloride is the dihydrochloride salt of α-fluoromethylhistidine, used as an irreversible inhibitor of histidine decarboxylase in biochemical research to reduce histamine levels. Supplied for research use only.

  • Irreversible inhibitor of histidine decarboxylase.
  • Reduces histamine levels in experimental models.
  • Supplied as the dihydrochloride salt to improve solubility and stability.
  • Molecular weight approximately 260.09 g/mol.
  • Purity reported at >98% by some suppliers.

Catalog No. 50-003-07649 Supplier Medchemexpress LLC Supplier No. HY1158221MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Fluoromethyl-L-histidine dihydrochloride is the dihydrochloride salt of α-fluoromethylhistidine, used as an irreversible inhibitor of histidine decarboxylase in biochemical research to reduce histamine levels. Supplied for research use only.

  • Irreversible inhibitor of histidine decarboxylase.
  • Reduces histamine levels in experimental models.
  • Supplied as the dihydrochloride salt to improve solubility and stability.
  • Molecular weight approximately 260.09 g/mol.
  • Purity reported at >98% by some suppliers.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.