Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical a-(Fmoc-amino)-3-tetrahydrofuran acetic acid | 100 g | CAS 1695476-98-2 | MDL MFCD28389395

Catalog No. 502353504
Encompass

a-(Fmoc-amino)-3-tetrahydrofuran acetic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1695476-98-2
- MDL: MFCD28389395
- InChIKey: BLVWTMAIPKJGEW-UHFFFAOYSA-N
- Molecular Weight: 367.401
- Molecular Formula: C21H21NO5
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29321900
- Country Of Origin: China
- IUPAC: 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-(tetrahydrofuran-3-yl)acetic acid
- SMILES: O=C(OCC1C2=CC=CC=C2C3=C1C=CC=C3)NC(C4CCOC4)C(O)=O

Catalog No. 50-235-3504 Supplier STA Pharmaceutical Supplier No. STAAA10180100GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

a-(Fmoc-amino)-3-tetrahydrofuran acetic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1695476-98-2
- MDL: MFCD28389395
- InChIKey: BLVWTMAIPKJGEW-UHFFFAOYSA-N
- Molecular Weight: 367.401
- Molecular Formula: C21H21NO5
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29321900
- Country Of Origin: China
- IUPAC: 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-(tetrahydrofuran-3-yl)acetic acid
- SMILES: O=C(OCC1C2=CC=CC=C2C3=C1C=CC=C3)NC(C4CCOC4)C(O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.