Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical a-(Fmoc-amino)-cyclobutaneacetic acid | 10 g | CAS 1695292-51-3 | MDL MFCD28389270

Catalog No. 502350447
Encompass

a-(Fmoc-amino)-cyclobutaneacetic acid is a Amino Acid reagent (Subcategory: Alkane AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1695292-51-3
- MDL: MFCD28389270
- InChIKey: TXLUWFPQLXODBP-UHFFFAOYSA-N
- Molecular Weight: 351.402
- Molecular Formula: C21H21NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclobutylacetic acid
- SMILES: O=C(OCC1C2=CC=CC=C2C3=C1C=CC=C3)NC(C4CCC4)C(O)=O

Catalog No. 50-235-0447 Supplier STA Pharmaceutical Supplier No. STAAA1028110GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

a-(Fmoc-amino)-cyclobutaneacetic acid is a Amino Acid reagent (Subcategory: Alkane AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1695292-51-3
- MDL: MFCD28389270
- InChIKey: TXLUWFPQLXODBP-UHFFFAOYSA-N
- Molecular Weight: 351.402
- Molecular Formula: C21H21NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclobutylacetic acid
- SMILES: O=C(OCC1C2=CC=CC=C2C3=C1C=CC=C3)NC(C4CCC4)C(O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.