Learn More
Medchemexpress LLC HY-102022 1mg , α-Galactosylceramide CAS:158021-47-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1020221MG
Encompass_Preferred
HY-102022 1mg α-Galactosylceramide CAS: 158021-47-7 α-Galactosylceramide (α-GalCer) is a synthetic glycolipid with antitumorial and immunostimulatory. α-Galactosylceramide is a very potent NKT cell agonist and binds effectively to CD1d. The complex of α-Galactosylceramide plus CD1d binds the NKT cell TCR (T cell antigen receptor)[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1254