Learn More
Cayman Chemical a-GunIdInoglutarIc AcId 100mg
SDP
Catalog No. 502894959
Encompass_Preferred
A linear mixed-type inhibitor of nNOS having a Ki value of 2.69 µM; normally present in small quantities in the cerebral cortex of cats with increased levels following seizure
Catalog No. 50-289-4959 Supplier Cayman Chemical Supplier No. 80340100 MG
A linear mixed-type inhibitor of nNOS having a Ki value of 2.69 µM; normally present in small quantities in the cerebral cortex of cats with increased levels following seizure