Learn More
Cayman Chemical a-GunIdInoglutarIc AcId 10mg
SDP
Supplier:  Cayman Chemical 8034010 MG
Encompass_Preferred
A linear mixed-type inhibitor of nNOS having a Ki value of 2.69 µM; normally present in small quantities in the cerebral cortex of cats with increased levels following seizure
Catalog No. 50-289-3931