Learn More
Cayman Chemical α-Helical CRF (9-41) (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502911382
Encompass_Preferred
A synthetic peptide CRF antagonist; inhibits CRF-induced ACTH release from isolated rat anterior pituitary cells from 0.5-5 µM; inhibits CRF-induced ACTH release in non-anesthetized, intact rats from 0.02-0.6 µmol/kg; inhibits stress-induced ACTH release in rats at 0.6 µmol/kg; increases social interaction time in a rat model of nicotine-induced conditioned anxiety; reduces binge-like ethanol consumption in mice
Catalog No. 50-291-1382 Supplier Cayman Chemical Supplier No. 3565010 MG
A synthetic peptide CRF antagonist; inhibits CRF-induced ACTH release from isolated rat anterior pituitary cells from 0.5-5 µM; inhibits CRF-induced ACTH release in non-anesthetized, intact rats from 0.02-0.6 µmol/kg; inhibits stress-induced ACTH release in rats at 0.6 µmol/kg; increases social interaction time in a rat model of nicotine-induced conditioned anxiety; reduces binge-like ethanol consumption in mice