Learn More
Bachem Pyr-AMC
SDP
Supplier:  Bachem 4004069.1000
Encompass_Preferred
Pyr-AMC, 66642-36-2, C15H14N2O4, Mr 286.29, 1g. A highly sensitive fluorogenic substrate for pyroglutamyl peptidase I.
Catalog No. 50-194-7073