Learn More
Bachem Fmoc-octyl-Gly-OH
SDP
Supplier:  Bachem 4062518.1000
Encompass_Preferred
Fmoc-octyl-Gly-OH, 193885-59-5, C25H31NO4, Mr 409.53, 1g. Fmoc-(S)-2-aminodecanoic acid, Fmoc-L-decyline, Fmoc-L-2-aminocapric acid. A homolg of Nle, lipophilic building block for SAR studies. A long-acting GLP-1 analog could be obtained by replacing Leu20 by L-octylglycine.
Catalog No. 50-194-7194