Learn More
Cayman Chemical a-hydroxy TamoxIfen 10mg
SDP
Catalog No. 502966957
Encompass_Preferred
A reactive metabolite of tamoxifen that is formed by the action of CYP3A4 in human liver; can be further converted into genotoxic DNA adducts though a mechanism involving reversible O-sulfonation of the hydroxyl moiety
Catalog No. 50-296-6957 Supplier Cayman Chemical Supplier No. 1720410 MG
A reactive metabolite of tamoxifen that is formed by the action of CYP3A4 in human liver; can be further converted into genotoxic DNA adducts though a mechanism involving reversible O-sulfonation of the hydroxyl moiety