Learn More
Cayman Chemical a-KaInIc AcIdhydrate 10mg
SDP
Catalog No. 502896450
Encompass_Preferred
A cyclic analog of L-glutamate; binds to GluK1, GluK2, GluK3, GluK4, and GluK5 (Kis = 75.9, 12.7, 32.8, 4.7, and 15 nM, respectively); induces calcium influx and membrane depolarization, as well as glutamate release, in rat brain synaptosomes at 5 mM; induces chromatin condensation and nuclear membrane fragmentation in primary neonatal cerebellar granule neurons at 100 µM; induces convulsive behavior in rats (ED50 = 0.51 nmol/animal) and induces seizures in mice (CD50 = 0.39 nmol/animal); has been commonly used to induce seizures in rodents
Catalog No. 50-289-6450 Supplier Cayman Chemical Supplier No. 7805010 MG
A cyclic analog of L-glutamate; binds to GluK1, GluK2, GluK3, GluK4, and GluK5 (Kis = 75.9, 12.7, 32.8, 4.7, and 15 nM, respectively); induces calcium influx and membrane depolarization, as well as glutamate release, in rat brain synaptosomes at 5 mM; induces chromatin condensation and nuclear membrane fragmentation in primary neonatal cerebellar granule neurons at 100 µM; induces convulsive behavior in rats (ED50 = 0.51 nmol/animal) and induces seizures in mice (CD50 = 0.39 nmol/animal); has been commonly used to induce seizures in rodents