Learn More
Cayman Chemical a-KetoglutarIc AcId 100g
SDP
Catalog No. 502946790
Encompass_Preferred
An intermediate in the citric acid cycle; precursor of glutamine and glutamate and an antioxidant with roles in immune homeostasis, aging, protein synthesis, and bone development
Catalog No. 50-294-6790 Supplier Cayman Chemical Supplier No. 33693100 G
An intermediate in the citric acid cycle; precursor of glutamine and glutamate and an antioxidant with roles in immune homeostasis, aging, protein synthesis, and bone development