Learn More
Cayman Chemical α-Ketoglutaric Acid, Purity: ≥95%, CAS: 328-50-7, 500 g
SDP
Catalog No. 502949973
Encompass_Preferred
An intermediate in the citric acid cycle; precursor of glutamine and glutamate and an antioxidant with roles in immune homeostasis, aging, protein synthesis, and bone development
Catalog No. 50-294-9973 Supplier Cayman Chemical Supplier No. 33693500 G
An intermediate in the citric acid cycle; precursor of glutamine and glutamate and an antioxidant with roles in immune homeostasis, aging, protein synthesis, and bone development