Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC A-ketoglutaric acid (disodium hydrate) | MFCD00150702 | 95.0% | 208.08 g/mol | C5H6Na2O6 | 500 MG
SDP

Catalog No. 5000280895
Encompass_Preferred

Alpha-ketoglutaric acid disodium hydrate is the disodium salt hydrate of alpha-ketoglutaric acid supplied as a white to off-white solid for laboratory research use. It serves as a metabolic intermediate and reagent for biochemical assays, enzymology, and method development. Documentation such as COA and SDS is available for quality and handling information.

  • Purity 95.0%.
  • Molecular formula C5H6Na2O6 and molecular weight 208.08 g/mol.
  • White to off-white solid appearance.
  • High water solubility: ~175 mg/mL (ultrasonication assisted).
  • Typical pack size 500 mg; larger sizes available on request.
  • Storage: long-term at 4°C under nitrogen; stock solutions at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-80895 Supplier Medchemexpress LLC Supplier No. HYW339545A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alpha-ketoglutaric acid disodium hydrate is the disodium salt hydrate of alpha-ketoglutaric acid supplied as a white to off-white solid for laboratory research use. It serves as a metabolic intermediate and reagent for biochemical assays, enzymology, and method development. Documentation such as COA and SDS is available for quality and handling information.

  • Purity 95.0%.
  • Molecular formula C5H6Na2O6 and molecular weight 208.08 g/mol.
  • White to off-white solid appearance.
  • High water solubility: ~175 mg/mL (ultrasonication assisted).
  • Typical pack size 500 mg; larger sizes available on request.
  • Storage: long-term at 4°C under nitrogen; stock solutions at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.