Learn More
Cayman Chemical α-Ketoglutaric Acid (sodium salt), Purity: ≥95%, CAS: 22202-68-2, 1 g
SDP
Catalog No. 502893728
Encompass_Preferred
An intermediate in the citric acid cycle; precursor of glutamine and glutamate and an antioxidant with roles in immune homeostasis, aging, protein synthesis, and bone development
Catalog No. 50-289-3728 Supplier Cayman Chemical Supplier No. 90018281 G
An intermediate in the citric acid cycle; precursor of glutamine and glutamate and an antioxidant with roles in immune homeostasis, aging, protein synthesis, and bone development