Learn More
Cayman Chemical a-KetoglutarIc AcIdodIum s 5g
SDP
Catalog No. 502894459
Encompass_Preferred
An intermediate in the citric acid cycle; precursor of glutamine and glutamate and an antioxidant with roles in immune homeostasis, aging, protein synthesis, and bone development
Catalog No. 50-289-4459 Supplier Cayman Chemical Supplier No. 90018285 G
An intermediate in the citric acid cycle; precursor of glutamine and glutamate and an antioxidant with roles in immune homeostasis, aging, protein synthesis, and bone development