Learn More
Bachem Fmoc-Suberol
SDP
Catalog No. 501947386
Encompass_Preferred
Fmoc-Suberol, 5-Fmoc-amino-2-carboxymethoxy-10,11-dihydro-5H-dibenzo[a,d]cycloheptene, 212783-75-0, C32H27NO5, Mr 505.57, 25g. Ramage Linker. A linker suitable for Fmoc-SPPS of peptide amides. Cleavage is achieved e.g. with 95% TFA, scavengers may be required.
Catalog No. 50-194-7386 Supplier Bachem Supplier No. 4025359.0025
Fmoc-Suberol, 5-Fmoc-amino-2-carboxymethoxy-10,11-dihydro-5H-dibenzo[a,d]cycloheptene, 212783-75-0, C32H27NO5, Mr 505.57, 25g. Ramage Linker. A linker suitable for Fmoc-SPPS of peptide amides. Cleavage is achieved e.g. with 95% TFA, scavengers may be required.