Learn More
Cayman Chemical a-LInolenIc AcId 100mg
SDP
Catalog No. 502892976
Encompass_Preferred
An essential ω-3 polyunsaturated fatty acid found in plants; converted to eicosapentaenoic, docosapentaenoic, and docosahexaenoic acids, catabolized via fatty acid β-oxidation for energy or the synthesis of saturated and monounsaturated fatty acids, or stored in adipose tissue in vivo; dietary consumption is positively associated with a moderately lower risk of cardiovascular disease
Catalog No. 50-289-2976 Supplier Cayman Chemical Supplier No. 90210100 MG
An essential ω-3 polyunsaturated fatty acid found in plants; converted to eicosapentaenoic, docosapentaenoic, and docosahexaenoic acids, catabolized via fatty acid β-oxidation for energy or the synthesis of saturated and monounsaturated fatty acids, or stored in adipose tissue in vivo; dietary consumption is positively associated with a moderately lower risk of cardiovascular disease