Learn More
Cayman Chemical a-LInolenIc AcId-d5 100UG
SDP
Catalog No. 502894282
Encompass_Preferred
An internal standard for the quantification of ALA by GC- or LC-MS
Catalog No. 50-289-4282 Supplier Cayman Chemical Supplier No. 9002286100 UG
An internal standard for the quantification of ALA by GC- or LC-MS