Learn More
Cayman Chemical a-LInolenoyl EthnolamIde 100mg
SDP
Catalog No. 502894838
Encompass_Preferred
An endocannabinoid containing α-linolenic acid in place of the arachidonate moiety of AEA; detected in porcine brain, but its specific role and relative importance as a cannabinergic neurotransmitter have not been elucidated
Catalog No. 50-289-4838 Supplier Cayman Chemical Supplier No. 90215100 MG
An endocannabinoid containing α-linolenic acid in place of the arachidonate moiety of AEA; detected in porcine brain, but its specific role and relative importance as a cannabinergic neurotransmitter have not been elucidated