Learn More
Cayman Chemical a-LInolenoyl EthnolamIde 10mg
SDP
Catalog No. 502892833
Encompass_Preferred
An endocannabinoid containing α-linolenic acid in place of the arachidonate moiety of AEA; detected in porcine brain, but its specific role and relative importance as a cannabinergic neurotransmitter have not been elucidated
Catalog No. 50-289-2833 Supplier Cayman Chemical Supplier No. 9021510 MG
An endocannabinoid containing α-linolenic acid in place of the arachidonate moiety of AEA; detected in porcine brain, but its specific role and relative importance as a cannabinergic neurotransmitter have not been elucidated