Learn More
Cayman Chemical α-Linolenoyl Ethanolamide, Purity: ≥98%, CAS: 57086-93-8, 50 mg
SDP
Catalog No. 502894839
Encompass_Preferred
An endocannabinoid containing α-linolenic acid in place of the arachidonate moiety of AEA; detected in porcine brain, but its specific role and relative importance as a cannabinergic neurotransmitter have not been elucidated
Catalog No. 50-289-4839 Supplier Cayman Chemical Supplier No. 9021550 MG
An endocannabinoid containing α-linolenic acid in place of the arachidonate moiety of AEA; detected in porcine brain, but its specific role and relative importance as a cannabinergic neurotransmitter have not been elucidated