Learn More
Bachem 3-Methoxytyramine · HCl
SDP
Supplier:  Bachem 4011849.0005
Encompass_Preferred
3-Methoxytyramine · HCl, 1477-68-5, C9H13NO2 · HCl, Mr 203.67, 5g. 3-MT · HCl, 3-O-Methyldopamine · HCl. A major metabolite of L-DOPA. Elevated concentrations of 3-MT in body fluids are indicators for a variety of brain and carcinoid tumors. N-Acetylation of 3-MT yields NAMDA, which in vitro inhibits GTP cyclohydrolase I, the rate-limiting enzyme for tetrahydrobiopterin biosynthesis. NAMDA also reduces nitrite accumulation, an oxidative metabolite of NO, without directly inhibiting NOS activity. CAS Number (net): 554-52-9.
Catalog No. 50-194-6736