Learn More
Bachem Polybia-MP1 Trifluoroacetate
SDP
Supplier:  Bachem 4099795.1000
Encompass_Preferred
Polybia-MP1 Trifluoroacetate, H-Ile-Asp-Trp-Lys-Lys-Leu-Leu-Asp-Ala-Ala-Lys-Gln-Ile-Leu-NH2, 872043-01-1, C78H132N20O19, Mr 1654.03, 1mg. A mastoparan-related antimicrobial insect venom peptide selectively inhibiting the proliferation of prostate and bladder cancer cells.
Catalog No. 50-194-7440