Learn More
Selleck Chemical LLC a-Methylbenzylamine
SDP
Catalog No. 5000003713
Encompass_Preferred
(S)-( )- -Methylbenzylamine is a chiral auxiliary in the enantiodivergent synthesis of simple isoquinoline alkaloids
Catalog No. 50-000-03713 Supplier Selleck Chemical LLC Supplier No. S626025UL
(S)-( )- -Methylbenzylamine is a chiral auxiliary in the enantiodivergent synthesis of simple isoquinoline alkaloids