Learn More
Medchemexpress LLC a-Methyltyrosine methyl ester (hydrochloride) | 7361-31-1 | 98.91% | 245.71 | 25 MG
SDP
Catalog No. 5000366651
Encompass_Preferred
α-Methyltyrosine methyl ester hydrochloride is an orally active and competitive tyrosine hydroxylase inhibitor. It inhibits the conversion of tyrosine to dopamine and can be used as a tool for sympathetic nervous system research. In rats, it has been observed to cause kidney damage and urethral calculi.
  • Orally active.
  • Competitive tyrosine hydroxylase inhibitor.
  • Inhibits the conversion of tyrosine to dopamine.
  • Can be used as a tool for sympathetic nervous system research.
Catalog No. 50-003-66651 Supplier Medchemexpress LLC Supplier No. 25MGHYW013407
α-Methyltyrosine methyl ester hydrochloride is an orally active and competitive tyrosine hydroxylase inhibitor. It inhibits the conversion of tyrosine to dopamine and can be used as a tool for sympathetic nervous system research. In rats, it has been observed to cause kidney damage and urethral calculi.
  • Orally active.
  • Competitive tyrosine hydroxylase inhibitor.
  • Inhibits the conversion of tyrosine to dopamine.
  • Can be used as a tool for sympathetic nervous system research.