Learn More
Electron Microscopy Sciences Ammonia Water 0.3% 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2639614
Encompass_Preferred
Used in Jones Method for Kidney, Silver Method For the staining of basement membranes, reticulum fibers, collagen and nuclei. References: Jones, D.B., Amer. J. Path. 27:99 (1951) AFIP Manual of Histological Staining Methods, 3rd Ed., Ed. L. Luna: NY: McGraw-Hill Publications, c. 1968, p.97
Catalog No. 50-318-10