Learn More
Bachem a-MSH Trifluoroacetate
SDP
Supplier:  Bachem 4008476.0005
Encompass_Preferred
a-MSH Trifluoroacetate, Ac-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-NH2, 581-05-5, C77H109N21O19S, Mr 1664.91, 5mg. Acetyl-ACTH (1-13) amide, a-Melanotropin (human). a-Melanotropin, also known as a-melanocyte-stimulating hormone (a-MSH), is a 13-amino acid peptide originally characterized as a neuropeptide derived from the pituitary gland. a-MSH is synthesized from pro-opiomelanocortin (POMC) by the action of specific prohormone convertases and is involved in the regulation of important physiological functions including food intake, energy homeostasis, modulation of immune responses and photoprotection.
Catalog No. 50-259-460