Learn More
Cayman Chemical a-MSH AcIdhumnmouserat 100mg
SDP
Catalog No. 502944554
Encompass_Preferred
An acid form of α-MSH; selectively binds to MC1R over MC3R, MC4R, and MC5R in COS-1 cells expressing the human receptors (Kis = 0.0012, 0.0829, 3.05, and 18.2 µM, respectively)
Catalog No. 50-294-4554 Supplier Cayman Chemical Supplier No. 41463100 MG
An acid form of α-MSH; selectively binds to MC1R over MC3R, MC4R, and MC5R in COS-1 cells expressing the human receptors (Kis = 0.0012, 0.0829, 3.05, and 18.2 µM, respectively)