Learn More
Apexbio Technology LLC a-MSH, amide, 5mg. Cas: MFCD: MFCD00133062. Melanocyte-stimulating hormones
SDP
Supplier:  Apexbio Technology LLC A10255
Encompass_Preferred
a-MSH, amide (C77H109N21O19S1), a peptide with the sequence Ac-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-NH2, MW= 1664.9. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4672