Learn More
American Radiolabeled Chemicals Inc A-MSH NDP 125I 10UC
SDP
Catalog No. 50901020
Encompass
a-MSH, Nle4, D-Phe7-125I 10uCi in buffered solution (pH 8.5), 0.075 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1770, Shipped in Dry Ice
Catalog No. 50-901-020 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 014210UC
a-MSH, Nle4, D-Phe7-125I 10uCi in buffered solution (pH 8.5), 0.075 mCi/ml, S.A. 2200 Ci/mmol, M.W. ~1770, Shipped in Dry Ice