Learn More
Cayman Chemical α-MSH (human, mouse, rat, porcine, bovine, ovine) (trifluoroacetate salt), Purity: ≥98%, CAS: 171869-93-5, 10 mg
SDP
Catalog No. 502988762
Encompass_Preferred
A peptide hormone; induces cAMP production in Hepa cells expressing human MC3R and MC4R (EC50s = 0.16 and 56 nM, respectively); reduces S. aureus colony formation and C. albicans germ tube formation in vitro at 100 pM; inhibits endotoxin-, ceramide-, TNF-α-, or okadaic acid-induced activation of NF-κB in U937 cells; reduces IL-6- or TNF-α-induced ear edema in mice; prevents the development of adjuvant-induced arthritis in rats and increases survival in a mouse model of septic shock; increased plasma levels are positively correlated with delayed disease progression and reduced death in patients with HIV
Catalog No. 50-298-8762 Supplier Cayman Chemical Supplier No. 2992310 MG
A peptide hormone; induces cAMP production in Hepa cells expressing human MC3R and MC4R (EC50s = 0.16 and 56 nM, respectively); reduces S. aureus colony formation and C. albicans germ tube formation in vitro at 100 pM; inhibits endotoxin-, ceramide-, TNF-α-, or okadaic acid-induced activation of NF-κB in U937 cells; reduces IL-6- or TNF-α-induced ear edema in mice; prevents the development of adjuvant-induced arthritis in rats and increases survival in a mouse model of septic shock; increased plasma levels are positively correlated with delayed disease progression and reduced death in patients with HIV