Learn More
Cayman Chemical α-Naphthoflavone, Purity: ≥98%, CAS: 604-59-1, 25 g
SDP
Catalog No. 502966470
Encompass_Preferred
A flavone that modulates xenobiotic metabolism; antagonizes AhR at nanomolar concentrations, although it agonizes AhR at higher concentrations (10 µM); inhibits CYP19, CYP1A1, CYP1A2, and CYP1B1 (IC50s = 500, 60, 6, and 5 nM, respectively), whereas it activates CYP3A4 (Kd = 7.4 µM)
Catalog No. 50-296-6470 Supplier Cayman Chemical Supplier No. 1692425 G
A flavone that modulates xenobiotic metabolism; antagonizes AhR at nanomolar concentrations, although it agonizes AhR at higher concentrations (10 µM); inhibits CYP19, CYP1A1, CYP1A2, and CYP1B1 (IC50s = 500, 60, 6, and 5 nM, respectively), whereas it activates CYP3A4 (Kd = 7.4 µM)