Learn More
World Precision Instrument A-P Microdrive,on Rail Clamp
SDP
Catalog No. 502539107
Encompass
The Parallel Rail Animal Stereotaxic Instrument provides superior performance. Up to six manipulator arms may be conveniently positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.
Catalog No. 50-253-9107 Supplier World Precision Instrument Supplier No. 505345
The Parallel Rail Animal Stereotaxic Instrument provides superior performance. Up to six manipulator arms may be conveniently positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.