Learn More
Sigma Aldrich Fine Chemicals Biosciences alpha-Phellandrene >=75%, stabilized | 99-83-2 | MFCD00040419 | 1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W2856091KGK
Encompass
alpha-Phellandrene >=75%, stabilized | Purity: >=75% | Mol Wt: 136.23 | 99-83-2 | MFCD00040419 | 1KG
Catalog No. NC1031193