Learn More
Bachem Pheromone Biosynthesis Activating Neuropeptide (Helicoverpa assulta, Heliothis zea)
SDP
Supplier:  Bachem 4039571.0500
Encompass_Preferred
Pheromone Biosynthesis Activating Neuropeptide (Helicoverpa assulta, Heliothis zea), H-Leu-Ser-Asp-Asp-Met-Pro-Ala-Thr-Pro-Ala-Asp-Gln-Glu-Met-Tyr-Arg-Gln-Asp-Pro-Glu-Gln-Ile-Asp-Ser-Arg-Thr-Lys-Tyr-Phe-Ser-Pro-Arg-Leu-NH2, 122071-54-9, C167H259N47O57S2, Mr 3901.31, 0.5mg. HeA-PBAN, HeZ-PBAN. A pheromone biosynthesis activating neuropeptide hormone which in the picomolar range induced sex pheromone synthesis in various moth species.
Catalog No. 50-194-7481